H26445-AA
H26445-AC
AAH26445-AAEA
24.1
USD
InStock
AAH26445-AA
AAH26445-AC
1-Bromo-2-ethylbutane 97%
1-Bromo-2-ethylbutane
Purity: 97%
CAS No.: 3814-34-4
Molecular Formula: (CH3CH2)2CHCH2Br
Formula Weight: 165.07
Storage Temperature: 2°C to 8°C
Flash Point: 36°(96°F)
Refractive Index: 1.4515
Odor: Sweet
Boiling Point: 145°
UN No.: 1993
MDL No.: MFCD00000219