H26743-03
H26743-06
AAH26743-03EA
55.5
USD
InStock
AAH26743-03
AAH26743-06
(S)-(-)-3-Amino-1,2-propanediol 98%
(S)-(-)-3-Amino-1,2-propanediol
Purity: 98%
CAS No.: 61278-21-5
Molecular Formula: C3H9NO2
Formula Weight: 91.11
Flash Point: 110°(230°F)
Refractive Index: 1.483
Melting Point: 54-56°
Boiling Point: 117-119/0.4mm
Beilstein Reference: 1719122
Optical Rotation: -28° (c=4, 5M HCl)
UN No.: 3259
MDL No.: MFCD00798260