Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-3-Bromo-α-methylbenzylamine ≥99%, ee 98%, ChiPros®

Danger

H26998-03 H26998-06
AAH26998-03EA 70.89 USD InStock
AAH26998-03 AAH26998-06
(S)-(-)-3-Bromo-α-methylbenzylamine ≥99%, ee 98%, ChiPros®
(S)-(-)-3-Bromo-α-methylbenzylamine
Purity: 99% CAS No.: 139305-96-7 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Boiling Point: 96°/4mm UN No.: 2735 MDL No.: MFCD00671638


Formula: C₈H₁₀BrN
Boiling Pt: 96 °C/4 mm
Density: 1.33
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671638
CAS Number: 139305-96-7
UN: 2735
ADR: "8",III
Order Now
Specification Test Results
Optical rotation -22° (neat)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty