Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Epoxyoctadecane tech. 85%, Technical Grade

Supplier: Origen Biomedical, Inc
Warning

L07622-14 L07622-22
AAAL07622-14EA 53.1 USD InStock
AAAL07622-14 AAAL07622-22
1,2-Epoxyoctadecane tech. 85%, Technical Grade
1,2-Epoxyoctadecane
Purity: 85% CAS No.: 7390-81-0 Molecular Formula: C18H36O Formula Weight: 268.49 Flash Point: 176°(348°F) Melting Point: 30-35° Boiling Point: 146-148°/1mm Beilstein Reference: 115434 MDL No.: MFCD00005152


Formula: C₁₈H₃₆O
Boiling Pt: 146…148 °C/1 mm
Melting Pt: 30…35 °C
Flash Pt: 176 °C(348 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005152
CAS Number: 7390-81-0
EINECS: 230-977-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty