Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetamidobenzoic acid ≥99.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

A0013-25G A0013-500G
TCA0013-025GEA 31 USD InStock
TCA0013-025G TCA0013-500G
4-Acetamidobenzoic acid ≥99.0% (by titrimetric analysis)
4-Acetamidobenzoic acid
CAS Number: 556-08-1
MDL Number: MFCD00002534
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 262


Formula: C₉H₉NO₃
MDL Number: MFCD00002534
CAS Number: 556-08-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d75da9d31b12448c3b95e63e69a640c722109b07&vid=d75da9d31b12448c3b95e63e69a640c722109b07&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty