Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoamyl acetate ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

A0033-500ML A0033-25ML
TCA0033-500MLEA 63.5 USD InStock
TCA0033-500ML TCA0033-25ML
Isoamyl acetate ≥98.0%
Isoamyl acetate
CAS Number: 123-92-2
MDL Number: MFCD00008946
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Flash Point (°C): 38
Specific Gravity (20/20): 0.87

Product Categories: Bioscience, Treatment Agents (Staining)
Other Product Names / Synonyms: Acetic Acid Isoamyl Ester, 3-Methylbutyl Acetate, Isopentyl Acetate


Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142
Melting Pt: -78
Density: 0.87 (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008946
CAS Number: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3 III
Merck Index: 12,05125
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f6b7448766aec298061b4cf89d8531a98e4e6f2&vid=4f6b7448766aec298061b4cf89d8531a98e4e6f2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty