A0034-500ML
A0034-25ML
TCA0034-500MLEA
37.18
USD
InStock
TCA0034-500ML
TCA0034-25ML
Isobutyl acetate ≥99.0%
Isobutyl acetate
CAS Number: 110-19-0
MDL Number: MFCD00008932
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 21
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Acetic Acid Isobutyl Ester