Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetone oxime ≥98.0%

Supplier: Decon Laboratories Ltd

A0065-25G A0065-500G
TCA0065-025GEA 23 USD InStock
TCA0065-025G TCA0065-500G
Acetone oxime ≥98.0%
Acetone oxime
CAS Number: 127-06-0
MDL Number: MFCD00002118
Molecular Formula: C3H7NO
Molecular Weight: 73.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 62


Formula: C₃H₇NO
Boiling Pt: 133 °C
Melting Pt: 62 °C
Density: 1.045 g/cm³ (20 °C)
MDL Number: MFCD00002118
CAS Number: 127-06-0
EINECS: 204-820-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,00077
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd4d42bdf1e7de2599d280905be572fa1fe5a744&vid=bd4d42bdf1e7de2599d280905be572fa1fe5a744&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty