Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-Aminopropyl)morpholine ≥99.0%

Supplier: Decon Laboratories Ltd

A0409-500ML A0409-25ML
TCA0409-500MLEA 224 USD InStock
TCA0409-500ML TCA0409-25ML
N-(3-Aminopropyl)morpholine ≥99.0%
N-(3-Aminopropyl)morpholine
CAS Number: 123-00-2
MDL Number: MFCD00006184
Molecular Formula: C7H16N2O
Molecular Weight: 144.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 105
Specific Gravity (20/20): 0.99

Other Product Names / Synonyms: 3-Morpholinopropylamine


Formula: C₇H₁₆N₂O
Boiling Pt: 225 °C
Density: 0.99 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00006184
CAS Number: 123-00-2
EINECS: 204-590-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e80f65e83a4f86bd982f28bbfbf1863caa9d733e&vid=e80f65e83a4f86bd982f28bbfbf1863caa9d733e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty