A0414-500G
A0414-25G
A0414-100G
TCA0414-500GEA
296
USD
InStock
TCA0414-500G
TCA0414-25G
TCA0414-100G
4-Aminopyridine ≥99.0%
4-Aminopyridine
CAS Number: 504-24-5
MDL Number: MFCD00006439
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 287
Melting point (°C): 158
Flash Point (°C): 164