A0891-500ML
A0891-25ML
TCA0891-500MLEA
360
USD
InStock
TCA0891-500ML
TCA0891-25ML
N-(3-Aminopropyl)cyclohexylamine ≥98.0% (by GC, titration analysis)
N-(3-Aminopropyl)cyclohexylamine
CAS Number: 3312-60-5
MDL Number: MFCD00003838
Molecular Formula: C9H20N2
Molecular Weight: 156.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 101
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: N-Cyclohexyl-1,3-propanediamine