Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminobenzyl alcohol ≥98.0%

Supplier: Decon Laboratories Ltd

A1000-25G A1000-250G
TCA1000-025GEA 94.2 USD InStock
TCA1000-025G TCA1000-250G
2-Aminobenzyl alcohol ≥98.0%
2-Aminobenzyl alcohol
CAS Number: 5344-90-1
MDL Number: MFCD00007749
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 85

Other Product Names / Synonyms: 2-Hydroxymethylaniline


Formula: C₇H₉NO
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c557b80b77f59a6d23a0c27dbe91897a2ef89ff&vid=0c557b80b77f59a6d23a0c27dbe91897a2ef89ff&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty