Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributylallyltin ≥95.0%

Supplier: Decon Laboratories Ltd
Danger

A1222-5G A1222-25G
TCA1222-005GEA 88 USD InStock
TCA1222-005G TCA1222-025G
Tributylallyltin ≥95.0%
Tributylallyltin
CAS Number: 24850-33-7
MDL Number: MFCD00010346
Molecular Formula: C15H32Sn
Molecular Weight: 331.13
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 103
Specific Gravity (20/20): 1.08


Formula: C₁₅H₃₂Sn
Boiling Pt: 155 °C
Density: 1.08 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty