Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trichlorobenzenesulfonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

B0247-500G B0247-25G
TCB0247-500GEA 727 USD InStock
TCB0247-500G TCB0247-25G
2,4,5-Trichlorobenzenesulfonyl chloride ≥98.0% (by titrimetric analysis)
2,4,5-Trichlorobenzenesulphonyl chloride
CAS Number: 15945-07-0
MDL Number: MFCD00007428
Molecular Formula: C6H2Cl4O2S
Molecular Weight: 279.94
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 68


Formula: C₆H₂Cl₄O₂S
Melting Pt: 68 °C
Density: 1.88 g/cm³ (20 °C)
MDL Number: MFCD00007428
CAS Number: 15945-07-0
EINECS: 240-079-0
UN: 3261
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2b3fb0cc95cf9b5972f55f712aa31aa41a7ad4a&vid=b2b3fb0cc95cf9b5972f55f712aa31aa41a7ad4a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty