B0522-500ML
B0522-25ML
B0522-100ML
TCB0522-500MLEA
41
USD
InStock
TCB0522-500ML
TCB0522-25ML
TCB0522-100ML
Trimethyl borate ≥98.0%
Trimethyl borate
CAS Number: 121-43-7
MDL Number: MFCD00008346
Molecular Formula: C3H9BO3
Molecular Weight: 103.91
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 68
Melting point (°C): -29
Flash Point (°C): -8
Specific Gravity (20/20): 0.93
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Boric Acid Trimethyl Ester, Methyl Borate