Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromopropionic acid ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

B0645-25G B0645-100G
TCB0645-25GEA 39.33 USD InStock
TCB0645-25G TCB0645-100G
3-Bromopropionic acid ≥98.0% (by GC, titration analysis)
3-Bromopropionic acid
CAS Number: 590-92-1
MDL Number: MFCD00002763
Molecular Formula: C3H5BrO2
Molecular Weight: 152.98
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 142
Melting point (°C): 63
Flash Point (°C): 66


Formula: BrH₂CCH₂COOH
Boiling Pt: 142 °C
Melting Pt: 63 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00002763
CAS Number: 590-92-1
EINECS: 209-694-1
UN: 3261
ADR: 8,II
Merck Index: 13,01418
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd19f12265976b3c7368e70449bf62249fcbebba&vid=fd19f12265976b3c7368e70449bf62249fcbebba&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty