Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Ethyl-2-bromopropionate ≥98.0%

Supplier: Decon Laboratories Ltd

B0646-500G B0646-25G B0646-100G
TCB0646-500GEA 146.5 USD InStock
TCB0646-500G TCB0646-25G TCB0646-100G
(±)-Ethyl-2-bromopropionate ≥98.0%
(±)-Ethyl-2-bromopropionate
CAS Number: 535-11-5
MDL Number: MFCD00000144
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 51
Specific Gravity (20/20): 1.40

Other Product Names / Synonyms: 2-Bromopropionic Acid Ethyl Ester


Formula: C₅H₉BrO₂
Boiling Pt: 160 °C
Density: 1.40 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-5
UN: 1992
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0ce7fa4a184a9a08a93c89296f12bc262cebd9b&vid=e0ce7fa4a184a9a08a93c89296f12bc262cebd9b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty