Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyramide ≥98.0% (by GC, total nitrogen)

Supplier: Decon Laboratories Ltd

B0809-25G B0809-500G
TCB0809-025GEA 25 USD InStock
TCB0809-025G TCB0809-500G
Butyramide ≥98.0% (by GC, total nitrogen)
Butyramide
CAS Number: 541-35-5
MDL Number: MFCD00041894
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 216
Melting point (°C): 115


Formula: C₄H₉NO
Boiling Pt: 216 °C
Melting Pt: 115 °C
Density: 1.027 g/cm³ (20 °C)
MDL Number: MFCD00041894
CAS Number: 541-35-5
EINECS: 208-776-4
Merck Index: 13,01592
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb9345907316893eef32fec656d8fab338dd8500&vid=cb9345907316893eef32fec656d8fab338dd8500&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty