Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-tert-Butylhydroquinone ≥98.0%

Supplier: Decon Laboratories Ltd

B0833-25G B0833-500G
TCB0833-025GEA 21.37 USD InStock
TCB0833-025G TCB0833-500G
2-tert-Butylhydroquinone ≥98.0%
2-tert-Butylhydroquinone
CAS Number: 1948-33-0
MDL Number: MFCD00002344
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 288
Melting point (°C): 127
Flash Point (°C): 161


Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 273
Melting Pt: 128
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00002344
CAS Number: 1948-33-0
EINECS: 217-752-2
UN: 3077
ADR: 9 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd2e733568880340ad3f538998c47e5759a50e34&vid=cd2e733568880340ad3f538998c47e5759a50e34&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty