Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltributylammonium bromide ≥98.0%

Supplier: Decon Laboratories Ltd

B1217-25G B1217-500G
TCB1217-025GEA 39 USD InStock
TCB1217-025G TCB1217-500G
Benzyltributylammonium bromide ≥98.0%
Benzyltributylammonium bromide
CAS Number: 25316-59-0
MDL Number: MFCD00011848
Molecular Formula: C19H34BrN
Molecular Weight: 356.39
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Other Product Names / Synonyms: Tributylbenzylammonium Bromide


Formula: C₁₉H₃₄BrN
Melting Pt: 174.5 - 175 °C
MDL Number: MFCD00011848
CAS Number: 25316-59-0
EINECS: 246-819-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3bcc75f53c2955eb06208314d3d37547c5748b6&vid=d3bcc75f53c2955eb06208314d3d37547c5748b6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty