Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S)-(+)-Camphor-10-sulphonic acid ≥98.0%

Supplier: Decon Laboratories Ltd

C0015-500G C0015-25G
TCC0015-500GEA 288.89 USD InStock
TCC0015-500G TCC0015-25G
(1S)-(+)-Camphor-10-sulphonic acid ≥98.0%
(1S)-(+)-Camphor-10-sulphonic acid
CAS Number: 3144-16-9
MDL Number: MFCD00064157
Molecular Formula: C10H16O4S
Molecular Weight: 232.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 200
Specific rotation [a]20/D: 21.5 deg (C=5, H2O)

Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Bioscience, Reagents for Oligosaccharide Synthesis, Bicyclic Monoterpenes
Other Product Names / Synonyms: (+)-CSA


Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Boiling Pt: 375
Melting Pt: 195
MDL Number: MFCD00064157
CAS Number: 3144-16-9
EINECS: 221-554-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b07c69f8dd1963b4d63f3af7f3469951296ca49&vid=5b07c69f8dd1963b4d63f3af7f3469951296ca49&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty