Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl carbonate ≥99.0%

Supplier: Decon Laboratories Ltd
Warning

C0043-25G C0043-500G
TCC0043-025GEA 23 USD InStock
TCC0043-025G TCC0043-500G
Diphenyl carbonate ≥99.0%
Diphenyl carbonate
CAS Number: 102-09-0
MDL Number: MFCD00003037
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 302
Melting point (°C): 80
Flash Point (°C): 168

Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Carbonic Acid Diphenyl Ester


Formula: C₁₃H₁₀O₃
Boiling Pt: 302 °C
Melting Pt: 80 °C
Density: 1.272 g/cm³ (14 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00003037
CAS Number: 102-09-0
EINECS: 203-005-8
Merck Index: 12,07433
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1cb0d00429f49103d09ec60adf65b815c427a8f&vid=d1cb0d00429f49103d09ec60adf65b815c427a8f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty