Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloroacetanilide ≥98.0%

Supplier: Decon Laboratories Ltd

C0089-25G C0089-500G
TCC0089-025GEA 57 USD InStock
TCC0089-025G TCC0089-500G
4'-Chloroacetanilide ≥98.0%
4'-Chloroacetanilide
CAS Number: 539-03-7
MDL Number: MFCD00000612
Molecular Formula: C8H8ClNO
Molecular Weight: 169.61
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 180


Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 333
Melting Pt: 180 °C
Density: 1.385 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000612
CAS Number: 539-03-7
EINECS: 208-707-8
Merck Index: 13,02128
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a017908d666ad13add7892b23b062609a35bbf7&vid=5a017908d666ad13add7892b23b062609a35bbf7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty