Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloroanilinium chloride ≥99.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd

C0113-500G C0113-25G
TCC0113-500GEA 171.6 USD InStock
TCC0113-500G TCC0113-25G
3-Chloroanilinium chloride ≥99.0% (by HPLC, total nitrogen)
3-Chloroanilinium chloride
CAS Number: 141-85-5
MDL Number: MFCD00035485
Molecular Formula: C6H6ClN
Molecular Weight: 164.03
Purity/Analysis Method: >99.0% (HPLC,N)
Form: Crystal


Formula: C₆H₇Cl₂N
MDL Number: MFCD00035485
CAS Number: 141-85-5
EINECS: 205-506-7
UN: 2018
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b83d55303827d7d6c3b655dac01577a97cf472&vid=28b83d55303827d7d6c3b655dac01577a97cf472&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty