Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroanilinium chloride ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

C0115-500G C0115-25G
TCC0115-500GEA 340 USD InStock
TCC0115-500G TCC0115-25G
4-Chloroanilinium chloride ≥98.0% (by titrimetric analysis)
4-Chloroanilinium chloride
CAS Number: 20265-96-7
MDL Number: MFCD00042057
Molecular Formula: C6H6ClN
Molecular Weight: 164.03
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 240
Lambda max.: 239 nm (H2O)


Formula: C₆H₇Cl₂N
Melting Pt: 240 °C
MDL Number: MFCD00042057
CAS Number: 20265-96-7
EINECS: 243-656-5
UN: 2018
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3729c4eb2f3089575fe7593c7898e0cfa8dd304&vid=f3729c4eb2f3089575fe7593c7898e0cfa8dd304&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty