Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloropentane ≥99.0%

Supplier: Decon Laboratories Ltd
Danger

C0237-500ML C0237-25ML
TCC0237-500MLEA 137 USD InStock
TCC0237-500ML TCC0237-25ML
1-Chloropentane ≥99.0%
1-Chloropentane
CAS Number: 543-59-9
MDL Number: MFCD00001015
Molecular Formula: C5H11Cl
Molecular Weight: 106.59
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 13
Specific Gravity (20/20): 0.88

Other Product Names / Synonyms: Amyl Chloride, Pentyl Chloride


Formula: C₅H₁₁Cl
Boiling Pt: 108 °C
Melting Pt: –99 °C
Density: 0.88 (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00001015
CAS Number: 543-59-9
EINECS: 208-846-4
UN: 1107
ADR: 3,II
Merck Index: 13,00610
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7732d8f30caadfceb281f3252eecd4eed5dc56c&vid=b7732d8f30caadfceb281f3252eecd4eed5dc56c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty