C0306-25ML
C0306-100ML
TCC0306-25MLEA
28.81
USD
InStock
TCC0306-25ML
TCC0306-100ML
Chlorotrimethylsilane ≥98.0%
Chlorotrimethylsilane
CAS Number: 75-77-4
MDL Number: MFCD00000502
Molecular Formula: C3H9ClSi
Molecular Weight: 108.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 57
Melting point (°C): -58
Flash Point (°C): -28
Specific Gravity (20/20): 0.86
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis, Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents, Bioscience, Reagents for Oligosaccharide Synthesis, Analytical Scien
Other Product Names / Synonyms: Trimethylchlorosilane, Trimethylsilyl Chloride, TMCS, TMS-Cl