Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Citrulline ≥98.0%

Supplier: Decon Laboratories Ltd

C0372-25G C0372-250G
TCC0372-025GEA 28.2 USD InStock
TCC0372-025G TCC0372-250G
L(+)-Citrulline ≥98.0%
L(+)-Citrulline
CAS Number: 372-75-8
MDL Number: MFCD00064397
Molecular Formula: C6H13N3O3
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 26 deg (C=8, 6mol/L HCl)


Formula: C₆H₁₃N₃O₃
MW: 175.19 g/mol
MDL Number: MFCD00064397
CAS Number: 372-75-8
EINECS: 206-759-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6868b177618982b929ec6ff5041a43fca8a656e1&vid=6868b177618982b929ec6ff5041a43fca8a656e1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty