Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Curcumin (natural)

Supplier: Decon Laboratories Ltd

C0434-1G C0434-25G
TCC0434-001GEA 24.42 USD InStock
TCC0434-001G TCC0434-025G
Curcumin (natural)
Curcumin
CAS Number: 458-37-7
MDL Number: MFCD00008365
Molecular Formula: C21H20O6
Molecular Weight: 368.39
Form: Crystal
Color: Pale Yellow
Melting point (°C): 183

Other Product Names / Synonyms: Natural Yellow 3, (E,E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione


Formula: C₂₁H₂₀O₆
Melting Pt: 183 °C
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fd9ede56e441b3df398508536acd00b40a6eb47&vid=4fd9ede56e441b3df398508536acd00b40a6eb47&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty