Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chlorophenyl)acetonitrile ≥98.0%

Supplier: Decon Laboratories Ltd

C0585-500G C0585-25G
TCC0585-500GEA 107 USD InStock
TCC0585-500G TCC0585-25G
(4-Chlorophenyl)acetonitrile ≥98.0%
(4-Chlorophenyl)acetonitrile
CAS Number: 140-53-4
MDL Number: MFCD00001918
Molecular Formula: C8H6ClN
Molecular Weight: 151.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 267
Flash Point (°C): 134
Freezing point (°C): 28

Other Product Names / Synonyms: (4-Chlorophenyl)acetonitrile


Formula: C₈H₆ClN
Boiling Pt: 267 °C
Melting Pt: 30.5 °C
Density: 1.1968 g/cm³ (20 °C)
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00001918
CAS Number: 140-53-4
EINECS: 205-418-9
UN: 3439
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c915b4925d872744420c4c8d7559b33d96acf65&vid=4c915b4925d872744420c4c8d7559b33d96acf65&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty