Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibromochloromethane ≥95.0% stabilized

Supplier: Decon Laboratories Ltd

C0854-5G C0854-25G
TCC0854-005GEA 62 USD InStock
TCC0854-005G TCC0854-025G
Dibromochloromethane ≥95.0% stabilized
Dibromochloromethane
(stabilized with Ethanol)
CAS Number: 124-48-1
MDL Number: MFCD00000820
Molecular Formula: CHBr2Cl
Molecular Weight: 208.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Freezing point (°C): -22
Specific Gravity (20/20): 2.42

Other Product Names / Synonyms: Chlorodibromomethane


Formula: Br₂ClCH
MDL Number: MFCD00000820
CAS Number: 124-48-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99312119282f4f5e418705bff82a1f2a1702d9b7&vid=99312119282f4f5e418705bff82a1f2a1702d9b7&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty