C1142-500G
C1142-25G
C1142-100G
TCC1142-500GEA
396.99
USD
InStock
TCC1142-500G
TCC1142-25G
TCC1142-100G
2-Chlorothanesulphonyl chloride ≥95.0% (by GC, titration analysis)
2-Chloroethanesulphonyl chloride
CAS Number: 1622-32-8
MDL Number: MFCD00007461
Molecular Formula: C2H4Cl2O2S
Molecular Weight: 163.01
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 203
Flash Point (°C): 152
Specific Gravity (20/20): 1.55
Storage Temperature: 0-10°C