Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Chlorobenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

C1270-25G C1270-100G
TCC1270-025GEA 156 USD InStock
TCC1270-025G TCC1270-100G
3-(4-Chlorobenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-(4-Chlorobenzoyl)propionic acid
CAS Number: 3984-34-7
MDL Number: MFCD00002794
Molecular Formula: C10H9ClO3
Molecular Weight: 212.63
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130


Formula: C₁₀H₉ClO₃
Melting Pt: 130 °C
MDL Number: MFCD00002794
CAS Number: 3984-34-7
EINECS: 223-627-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2048ed2d5c5a85cef09d87863e0d06020400e45&vid=d2048ed2d5c5a85cef09d87863e0d06020400e45&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty