Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-p-toluonitrile ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

C1398-250G C1398-25G
TCC1398-250GEA 466 USD InStock
TCC1398-250G TCC1398-25G
α-Bromo-p-toluonitrile ≥98.0% (by GC, titration analysis)
α-Bromo-p-toluonitrile
CAS Number: 17201-43-3
MDL Number: MFCD00001829
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 116

Other Product Names / Synonyms: 4-(Bromomethyl)benzonitrile, alpha-Bromo-p-tolunitrile


Formula: C₈H₆BrN
Boiling Pt: 143 °C
Melting Pt: 116 °C
MDL Number: MFCD00001829
CAS Number: 17201-43-3
EINECS: 241-246-0
UN: 2923
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7488db8aae19be482ac24cdb5edb21afb678aa1&vid=a7488db8aae19be482ac24cdb5edb21afb678aa1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty