C1617-500ML
C1617-25ML
TCC1617-500MLEA
123.31
USD
InStock
TCC1617-500ML
TCC1617-25ML
Cyclopentene ≥95.0%
Cyclopentene
CAS Number: 142-29-0
MDL Number: MFCD00001394
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 46
Melting point (°C): -135
Flash Point (°C): -29
Specific Gravity (20/20): 0.77
Storage Temperature: 0-10°C