Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzylamine ≥97.0%

Supplier: Decon Laboratories Ltd
Warning

D0152-25G D0152-500G
TCD0152-025GEA 31 USD InStock
TCD0152-025G TCD0152-500G
Dibenzylamine ≥97.0%
Dibenzylamine
CAS Number: 103-49-1
MDL Number: MFCD00004770
Molecular Formula: C14H15N
Molecular Weight: 197.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 270
Melting point (°C): -26
Flash Point (°C): 150
Specific Gravity (20/20): 1.03


Formula: C₆H₅CH₂NHCH₂C₆H₅
MW: 197.28 g/mol
Boiling Pt: 300
Melting Pt: -26
Density: 1.03 (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004770
CAS Number: 103-49-1
EINECS: 203-117-7
UN: 2810
ADR: 6.1 III
Merck Index: 13,03035
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45904366b8ca909d91a366def76ade7c88380370&vid=45904366b8ca909d91a366def76ade7c88380370&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty