Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Dichlorohexane ≥98.0%

Supplier: Decon Laboratories Ltd

D0372-500G D0372-25G
TCD0372-500GEA 84.7 USD InStock
TCD0372-500G TCD0372-25G
1,6-Dichlorohexane ≥98.0%
1,6-Dichlorohexane
CAS Number: 2163-00-0
MDL Number: MFCD00001020
Molecular Formula: C6H12Cl2
Molecular Weight: 155.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 74
Specific Gravity (20/20): 1.07

Other Product Names / Synonyms: Hexamethylene Chloride, Hexamethylene Dichloride


Formula: ClH₂C(CH₂)₄CH₂Cl
Boiling Pt: 95 °C
Melting Pt: –13 °C
Density: 1.07 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001020
CAS Number: 2163-00-0
EINECS: 218-491-7
UN: 3082
ADR: 9,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4068ac339d137d085040bbe5de524771158cc9aa&vid=4068ac339d137d085040bbe5de524771158cc9aa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty