Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloromethylvinylsilane ≥97.0%

Supplier: Decon Laboratories Ltd

D0378-500ML D0378-25ML
TCD0378-500MLEA 185 USD InStock
TCD0378-500ML TCD0378-025ML
Dichloromethylvinylsilane ≥97.0%
Dichloromethylvinylsilane
CAS Number: 124-70-9
MDL Number: MFCD00000492
Molecular Formula: C3H6Cl2Si
Molecular Weight: 141.07
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 14
Specific Gravity (20/20): 1.09


Formula: C₃H₆Cl₂Si
MW: 141.07 g/mol
Boiling Pt: 94 °C
Density: 1.09 (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00000492
CAS Number: 124-70-9
EINECS: 204-710-3
UN: 2985
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a521e930bb92c3da4d65e9cef30dc4c822a7217&vid=9a521e930bb92c3da4d65e9cef30dc4c822a7217&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty