Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisobutylamine

Supplier: Decon Laboratories Ltd

D0614-500ML D0614-25ML
TCD0614-500MLEA 41.7 USD InStock
TCD0614-500ML TCD0614-25ML
Diisobutylamine
Diisobutylamine
CAS Number: 110-96-3
MDL Number: MFCD00008930
Molecular Formula: C8H19N
Molecular Weight: 129.25
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 139
Melting point (°C): -74
Flash Point (°C): 29
Specific Gravity (20/20): 0.75


Formula: C₈H₁₉N
Boiling Pt: 139 °C
Melting Pt: –74 °C
Density: 0.75 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00008930
CAS Number: 110-96-3
EINECS: 203-819-3
UN: 2361
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=346563a3e4f711a2191452fb6eaeebcc6d4f652f&vid=346563a3e4f711a2191452fb6eaeebcc6d4f652f&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty