Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethylpentane ≥99.0%

Supplier: Decon Laboratories Ltd
Danger

D0769-5ML D0769-25ML
TCD0769-005MLEA 71.1 USD InStock
TCD0769-005ML TCD0769-25ML
2,4-Dimethylpentane ≥99.0%
2,4-Dimethylpentane
CAS Number: 108-08-7
MDL Number: MFCD00008945
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): -12
Specific Gravity (20/20): 0.67

Other Product Names / Synonyms: Diisopropylmethane


Formula: C₇H₁₆
MW: 100.2 g/mol
Boiling Pt: 81
Melting Pt: -119.2
Density: 0.67 (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008945
CAS Number: 108-08-7
EINECS: 203-548-0
UN: 1206
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6452da0f56a7727d406e575ccd27a57d93a9cb9a&vid=6452da0f56a7727d406e575ccd27a57d93a9cb9a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty