Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Xylenol ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

D0774-500G D0774-25G
TCD0774-500GEA 105 USD InStock
TCD0774-500G TCD0774-25G
2,4-Xylenol ≥98.0%
2,4-Xylenol
CAS Number: 105-67-9
MDL Number: MFCD00002233
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 112
Specific Gravity (20/20): 1.02

Other Product Names / Synonyms: 2,4-Xylenol


Formula: C₈H₁₀O
Boiling Pt: 212 °C
Melting Pt: 25 °C
Density: 1.02 (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b2173f97cf3fcfd12b66fff278fe903dd572a5&vid=85b2173f97cf3fcfd12b66fff278fe903dd572a5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty