Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl-m-toluidine ≥97.0%

Supplier: Decon Laboratories Ltd
Danger

D0806-500ML D0806-25ML
TCD0806-500MLEA 141 USD InStock
TCD0806-500ML TCD0806-25ML
N,N-Dimethyl-m-toluidine ≥97.0%
N,N-Dimethyl-m-toluidine
CAS Number: 121-72-2
MDL Number: MFCD00008305
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 85
Specific Gravity (20/20): 0.94

Other Product Names / Synonyms: N,N,3-Trimethylaniline, 3-Dimethylaminotoluene


Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 209
Melting Pt: -15
Density: 0.94 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00008305
CAS Number: 121-72-2
EINECS: 204-495-6
UN: 1708
ADR: 6.1 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78b1ce54b4d339154aa7c8f4f00e64e831c11b46&vid=78b1ce54b4d339154aa7c8f4f00e64e831c11b46&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty