Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dibutyltrimethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D1078-500ML D1078-25ML
TCD1078-500MLEA 219 USD InStock
TCD1078-500ML TCD1078-25ML
N,N-Dibutyltrimethylenediamine ≥98.0% (by GC, titration analysis)
N,N-Dibutyltrimethylenediamine
CAS Number: 102-83-0
MDL Number: MFCD00008219
Molecular Formula: C11H26N2
Molecular Weight: 186.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 104
Specific Gravity (20/20): 0.83

Other Product Names / Synonyms: N,N-Dibutyl-1,3-diaminopropane, N,N-Dibutyl-1,3-propanediamine, N,N-Dibutyltrimethylenediamine


Formula: C₁₁H₂₆N₂
MDL Number: MFCD00008219
CAS Number: 102-83-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee09efc996e67e6852482c9723c997b644d31fe4&vid=ee09efc996e67e6852482c9723c997b644d31fe4&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty