Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlorosalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D1112-25G D1112-500G
TCD1112-025GEA 46 USD InStock
TCD1112-025G TCD1112-500G
3,5-Dichlorosalicylic acid ≥98.0% (by GC, titration analysis)
3,5-Dichlorosalicylic acid
CAS Number: 320-72-9
MDL Number: MFCD00002442
Molecular Formula: C7H4Cl2O3
Molecular Weight: 207.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 227

Other Product Names / Synonyms: 3,5-Dichloro-2-hydroxybenzoic Acid


Formula: C₇H₄Cl₂O₃
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 227 °C
MDL Number: MFCD00002442
CAS Number: 320-72-9
EINECS: 206-281-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a0ae53487739b58f016255c682d1af6599ee83f&vid=7a0ae53487739b58f016255c682d1af6599ee83f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty