Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥98.0%

Supplier: Decon Laboratories Ltd

D1691-25G D1691-5G D1691-1G
TCD1691-25GEA 311 USD InStock
TCD1691-25G TCD1691-005G TCD1691-001G
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥98.0%
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
CAS Number: 22323-82-6
MDL Number: MFCD00063239
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 80
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: 14 deg (neat)

Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(+)-1,2-Isopropylideneglycerol


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 82
Density: 1.07 (20 °C)
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063239
CAS Number: 22323-82-6
EINECS: 244-910-8
Merck Index: 12,05232
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d669c8829cf90ec8d4024aa7ed24d5cdc79a61&vid=40d669c8829cf90ec8d4024aa7ed24d5cdc79a61&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty