Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimidium bromide ≥98.0%

Supplier: Decon Laboratories Ltd
Warning

D1815-5G D1815-1G
TCD1815-5GEA 973.77 USD InStock
TCD1815-5G TCD1815-001G
Dimidium bromide ≥98.0%
Dimidium bromide
CAS Number: 518-67-2
MDL Number: MFCD00011757
Molecular Formula: C20H18BrN3
Molecular Weight: 380.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Product Categories: Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3,8-Diamino-5-methyl-6-phenylphenanthridinium Bromide


Formula: C₂₀H₁₈BrN₃
MW: 380.29 g/mol
Melting Pt: 243…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011757
CAS Number: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1 I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=524298236bb416a7574b2e12a8bf000d52b317ca&vid=524298236bb416a7574b2e12a8bf000d52b317ca&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty