Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Digoxin ≥96.0% (by HPLC)

Supplier: Decon Laboratories Ltd

D1828-100MG D1828-1G
TCD1828-100MGEA 30.53 USD InStock
TCD1828-100MG TCD1828-1G
Digoxin ≥96.0% (by HPLC)
Digoxin
CAS Number: 20830-75-5
MDL Number: MFCD00003674
Molecular Formula: C41H64O14
Molecular Weight: 780.95
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 265
Specific rotation [a]20/D: 12 deg (C=10, Pyridine)


Formula: C₄₁H₆₄O₁₄
MDL Number: MFCD00003674
CAS Number: 20830-75-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39bdc04f8f1727c1ff674d7a96509ef4ea083c6e&vid=39bdc04f8f1727c1ff674d7a96509ef4ea083c6e&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty