Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethoxyaniline ≥98.0%

Supplier: Decon Laboratories Ltd

D1893-500G D1893-25G
TCD1893-500GEA 134 USD InStock
TCD1893-500G TCD1893-25G
2,5-Dimethoxyaniline ≥98.0%
2,5-Dimethoxyaniline
CAS Number: 102-56-7
MDL Number: MFCD00008368
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 81
Flash Point (°C): 150

Other Product Names / Synonyms: Aminohydroquinone Dimethyl Ether


Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 270
Melting Pt: 81
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008368
CAS Number: 102-56-7
EINECS: 203-040-9
UN: 2811
ADR: 6.1 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d76b81a354daa3c0249fc0c011c3e5b8cd349b0&vid=4d76b81a354daa3c0249fc0c011c3e5b8cd349b0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty