Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenetole ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

E0043-500ML E0043-25ML
TCE0043-500MLEA 61 USD InStock
TCE0043-500ML TCE0043-025ML
Phenetole ≥98.0%
Phenetole
CAS Number: 103-73-1
MDL Number: MFCD00009090
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Melting point (°C): -30
Flash Point (°C): 57
Specific Gravity (20/20): 0.97

Other Product Names / Synonyms: Ethoxybenzene, Ethyl Phenyl Ether


Formula: C₆H₅OC₂H₅
Boiling Pt: 173 °C
Melting Pt: –30 °C
Density: 0.97 (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00009090
CAS Number: 103-73-1
EINECS: 203-139-7
UN: 3271
ADR: 3,III
Merck Index: 12,07374
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f68d46a525d051512916ee99cca86c0fd407817&vid=4f68d46a525d051512916ee99cca86c0fd407817&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty