Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylmorpholine ≥99.0%

Supplier: Decon Laboratories Ltd

E0145-500ML E0145-25ML
TCE0145-500MLEA 48 USD InStock
TCE0145-500ML TCE0145-25ML
4-Ethylmorpholine ≥99.0%
4-Ethylmorpholine
CAS Number: 100-74-3
MDL Number: MFCD00006177
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 139
Melting point (°C): -63
Flash Point (°C): 36
Specific Gravity (20/20): 0.91


Formula: C₆H₁₃NO
Boiling Pt: 139 °C
Melting Pt: –63 °C
Density: 0.91 (20 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00006177
CAS Number: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8be9bbec17db5186eda23126af37477e1223d9a&vid=c8be9bbec17db5186eda23126af37477e1223d9a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty